Nitrile Examination Gloves, Tronex®
: Tronex
Offering superior barrier protection, advanced nitrile formulation offers tensile strength, stretch, and fit.
- 100% protein free
- Textured surface for secure grip
- Sterilized with gamma radiation at minimum 25 kGy
- 2" turned-up cuff and inner wrap in 'Easy-Peel' pouch to maintain sterile integrity
: Packaged in a convenient easy-peel pouch in a dispenser box.
- Catalog No:
- 10153-006
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable/Reusable:
- Disposable
- Latex/Latex-free:
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- ASTM No.:
- D6319
Frequently Bought Together