To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Semaphorin Cytoplasmic Domain Associated Protein 3
- Antigen Symbol:SEMCAP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:23024
- Antigen Synonyms:likely ortholog of mouse semaF cytoplasmic domain associated protein 3|PDZ domain containing ring finger 3|LNX3|SEMACAP3|Protein SEMACAP3|Ligand of Numb protein X 3|PZRN3_HUMAN.|Semaphorin cytoplasmic domain associated protein
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10668-800
- Supplier no.:BS-9191R
Specifications
About this item
PDZRN3 contains a RING-finger motif in its N-terminal region, two PDZ domains in its central region and a consensus-binding motif for PDZ domains at its C-terminus. It was identified in silico as a homolog of the protein known as LNX1 or SEMCAP1, which possesses ubiquitin ligase activity and binds the membrane protein Semaphorin 4C. However, PDZRN3 itself has not previously been characterized. We have now evaluated the properties and functions of PDZRN3. The PDZRN3 gene was shown to be expressed in various human tissues including the heart, skeletal muscle and liver and its expression in mouse skeletal muscle was developmentally regulated. Both the differentiation of C2C12 mouse skeletal myoblasts into myotubes and injury-induced muscle regeneration in vivo were found to be accompanied by up-regulation of PDZRN3. The differentiation-associated increase in the expression of PDZRN3 in C2C12 cells follows that of myogenin and precedes that of myosin heavy chain. Depletion of PDZRN3 by RNA interference inhibited the formation of myotubes as well as the associated up-regulation of myosin heavy chain in C2C12 cells. Our data suggest that PDZRN3 plays an essential role in the differentiation of myoblasts into myotubes by acting either downstream or independently of myogenin.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: SEMCAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat