Anti-MAP4K4 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
MAP4K4 (Mitogen-activated protein kinase kinase kinase kinase 4) belongs to the serine/threonine protein kinase family and specifically activates MAPK8/JNK. It may play a role in the response to environmental stress and is thought to function through the MAP3K7-MAP2K4-MAP2K7 kinase cascade, and mediate the TNF alpha signaling pathway. It interacts with the SH3 domain of the adapter protein Nck. MAP4K4-dependent signaling inhibits PPARG responsive gene expression, adipogenesis, and insulin stimulated glucose transport.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: MAP4K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-5492R
Datasheet
Specifications
- Size:100 µg
- Cross Adsorption:No
- Clonality:Polyclonal
- Modification:Ser631
- Purification:Purified by Protein A
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Isotype:IgG
- Host:Rabbit
- Concentration:1 μg/μl
- Cat. No.:10418-546
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:4°C
- Antibody Type:Primary
- Antigen Symbol:MAP4K4
- Gene ID:9448
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:MEKKK4|HEL-S-31|HGK|FLH21957|NIK
- Antigen Name:Mitogen-activated Protein kinase kinase kinase kinase 4
- Storage Temperature:Store at –20 °C for 12 months
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



