Anti-G gamma14 Rabbit Polyclonal Antibody
Catalog # 10293-852
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:G gamma 8
- Antigen Symbol:GNG8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:guanine nucleotide binding protein gamma 9|SG|Ogamma T2 subunit precursor|G gamma 9|IG|gamma transducing|GBGT2_HUMAN.|guanine nucleotide binding protein gamma transducing activity|GNG8|GNG9|G GAMMA C|GNGT2|G GAMMA 8|GNGT8|G protein cone gamma 8 subunit|guanine nucleotide binding protein G protein|Guanine nucleotide binding protein G|gamma T2 subunit
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10293-852
- Supplier no.:BS-13243R
Specifications
About this item
G Gamma14 (GNGT2) is a Guanine nucleotide-binding protein. G proteins are composed of 3 units, alpha, beta and gamma and are involved in various transmembrane signaling systems as signal modulators or transducers. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction. This G protein is specific to retinal cones and is involved in regulation of phototransduction.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: G gamma14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse