Anti-KLC1/KNS2 Rabbit Polyclonal Antibody
Catalog # 10251-430
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Symbol:KLC1/KNS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:liquid
- Gene ID:3831
- Antigen Synonyms:KNS2|KLC 1|Kinesin light chain 1|KLC|KLC1|KNS2A
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:1-50/573
- Purification:Purified by Protein A
- Cat. No.:10251-430
- Supplier no.:BS-11212R
Specifications
About this item
Kinesin is a microtubule-associated force-producing protein that may play a role in organelle transport. The light chain may function in coupling of cargo to the heavy chain or in the modulation of its ATPase activity.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: KLC1/KNS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat