Anti-ZFP103 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
10487-352
: Bioss
- Antibody Type:Primary
- Antigen Name:Zinc Finger Protein 103
- Antigen Symbol:ZFP103
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:7844
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10487-352
- Supplier no.:BS-9382R-HRP
Acts as an E2-dependent E3 ubiquitin-protein ligase, probably involved in the ER-associated protein degradation pathway.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: ZFP103
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat