Anti-TTC33 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # 10667-976
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Tetratricopeptide Repeat Doman 33
- Antigen Symbol:TTC33
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:23548
- Antigen Synonyms:OSRF|TTC33|TPR repeat protein 33|Tetratricopeptide repeat protein 33|Osmosis-responsive factor
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:201-250/262
- Purification:Purified by Protein A
- Cat. No.:10667-976
- Supplier no.:BS-9146R-A647
Specifications
About this item
TTC33 contains 3 TPR repeats. The function of the TTC33 protein remains unknown.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TTC33
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat