Anti-CD28 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10664-924
: Bioss
- Antibody Type:Primary
- Antigen Name:CD28 Molecule
- Antigen Symbol:CD28
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:940
- Antigen Synonyms:Tp44|MGC138290|T-cell-specific surface glycoprotein CD28|CD28 antigen
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:19-69/220
- Purification:Purified by Protein A
- Cat. No.:10664-924
- Supplier no.:BS-8865R-FITC
Involved in T-cell activation, the induction of cell proliferation and cytokine production and promotion of T-cell survival.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CD28
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat