Specifications
- Antibody Type:Primary
- Antigen Name:Xanthine Dehydrogenase
- Antigen Symbol:XDH
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:7498
- Antigen Synonyms:xanthine dehydrogenase|XOR|XO
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10662-924
- Supplier no.:BS-8552R-A555
Specifications
About this item
Key enzyme in purine degradation. Catalyzes the oxidation of hypoxanthine to xanthine. Catalyzes the oxidation of xanthine to uric acid. Contributes to the generation of reactive oxygen species. Has also low oxidase activity towards aldehydes (in vitro).Involvement in disease;Defects in XDH are the cause of xanthinuria type 1 (XU1) . Xanthinuria is characterized by excretion of very large amounts of xanthine in the urine and a tendency to form xanthine stones. Uric acid is strikingly diminished in serum and urine. XU1 is due to isolated xanthine dehydrogenase. XU1 patients can metabolize allopurinol.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: XDH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat