Anti-SLU7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10481-958
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:SLU7 Homolog, Splicing Factor
- Antigen Symbol:SLU7
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:10569
- Antigen Synonyms:hSlu7|SLU7 splicing factor homolog S. cerevisiae|9G8
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10481-958
- Supplier no.:BS-8327R-FITC
Specifications
About this item
Participates in the second catalytic step of pre-mRNA splicing, when the free hydroxyl group of exon I attacks the 3'-splice site to generate spliced mRNA and the excised lariat intron. Required for holding exon 1 properly in the spliceosome and for correct AG identification when more than one possible AG exists in 3'-splicing site region. May be involved in the activation of proximal AG. Probably also involved in alternative splicing regulation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLU7
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat