Anti-KLHL24 Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10475-618
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Kelch-like 24
- Antigen Symbol:KLHL24
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:54800
- Antigen Synonyms:KRIP6|DRE1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:15-60/600
- Purification:Purified by Protein A
- Cat. No.:10475-618
- Supplier no.:BS-8050R-CY5
Specifications
About this item
Specifically reduces kainate receptor-mediated currents in hippocampal neurons, most probably by modulating channel properties.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KLHL24
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat