To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Hippocampus Abundant Transcript 1
- Antigen Symbol:HIAT1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:64645
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10475-462
- Supplier no.:BS-8042R-CY3
The Major facilitator superfamily consists of presumed carbohydrate transporters with 10-12 membrane-spanning domains. Belonging to the facilitator superfamily, HIAT1 is a 490 amino acid multi-pass membrane protein that may function as a sugar transporter and is expressed in adult and embryonic brain. The HIAT1 gene was first observed while analyzing for active genes in neonatal mouse hippocampus. The gene encoding HIAT1 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. Stickler syndrome, Parkinsons, schizophrenia, familial adenomatous polyposis, Gaucher disease and Usher syndrome are also associated with chromosome 1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HIAT1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat