Anti-SRPK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10472-868
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:SRSF Protein Kinase 2
- Antigen Symbol:SRPK2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:6733
- Antigen Synonyms:SR protein specic kinase 2|Serine kinase SRPK2|SRPK2_HUMAN.|Human serine kinase SRPK2 mRNA|complete cds|Serine/threonine protein kinase SRPK2|SFRSK2|SFRS protein kinase 2|Serine/arginine rich protein specic kinase 2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10472-868
- Supplier no.:BS-7923R-FITC
Specifications
About this item
SRPK2 belongs to the protein kinase superfamily. It phosphorylates RS domain-containing proteins, such as SFRS1 and SFRS2 on serine residues. It has a role in spliceosome assembly and in mediating the trafficking of splicing factors and appears to mediate HBV core protein phosphorylation which is a prerequisite for pregenomic RNA encapsidation into viral capsids. SRPK2 highly expressed in brain, moderately expressed in heart and skeletal muscle and at low levels in lung, liver, and kidney.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SRPK2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat