Order Entry
United States
ContactUsLinkComponent
 
Anti-PCBP4 Rabbit Polyclonal Antibody (Cy7®)
  10465-892
 :  Bioss
Anti-PCBP4 Rabbit Polyclonal Antibody (Cy7®)
  10465-892
 :  Bioss
 :  BS-7616R-CY7

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Poly(rC) Binding Protein 4
  • Antigen Symbol:
    PCBP4
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    57060
  • Antigen Synonyms:
    LIP4|LYST-interacting protein|PCBP4|poly(rC) binding protein 4 |alphaCP-4 protein|alpha CP-4|poly(rC)-binding protein 4|MCG10 |RNA binding protein MCG10
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10465-892
  • Supplier no.:
    BS-7616R-CY7

 

 

MCG10 is a member of the KH-domain RNA-binding protein family. Proteins of this family bind to RNA with specificity for C-rich pyrimidine regions. MCG10 is induced by p53 and DNA damage via two p53-responsive promoter elements.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCBP4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat