Anti-EI24 Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10451-582
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:etoposide induced 2.4
- Antigen Symbol:EI24
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:9538
- Antigen Synonyms:EPG4|TP53I8|PIG8
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10451-582
- Supplier no.:BS-6637R-CY3
Specifications
About this item
This gene has higher expression in p53-expressing cells than in control cells and is an immediate-early induction target of p53-mediated apoptosis. The protein encoded by this gene contains six putative transmembrane domains and may suppress cell growth by inducing apoptotic cell death through the caspase 9 and mitochondrial pathways. This gene is located on human chromosome 11q24, a region frequently altered in cancers. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EI24
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat