To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cathelicidin
- Antigen Symbol:CAMP
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:820
- Antigen Synonyms:FALL-39|CAP18|CAP-18|LL37|CRAMP|Cathelicidin antimicrobial peptide|18 kDa cationic antimicrobial protein|HSD26|FALL39
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10447-000
- Supplier no.:BS-6433R-FITC
Specifications
About this item
Cathelicidins are a family of antimicrobial proteins found in the peroxidase-negative granules of neutrophils. Along with the family of proteins known as defensins, cathelicidins participate in the first line of defense by preventing local infection and systemic invasion of microbes. FALL-39 precursor (FALL-39 peptide antibiotic, cationic anti-microbial protein, CAMP, CAP-18, HSD26) is a cathelicidin anti-microbial protein that contains the antibacterial peptide LL-37 (amino acids 134-170). In contrast to the defensins, which are cysteine-rich peptides that fold in ∫-pleated sheets, LL-37 is a cysteine-free peptide that can adopt an amphipathic å-helical conformation. LL-37 binds to bacterial lipopolysaccharides (LPS) and is a potent chemotactic factor for recruiting mast cells to sites of inflammation. LL-37 is present in inflammatory skin diseases that include psoriasis, sub-acute lupus erthematosus, dermatitis and nickel contact hypersensitivity. It is not found in normal skin epidermis. The secreted protein is expressed primarily in bone marrow, testis and neutrophils. The mouse and rat ortholog, CRAMP (cathelin-related antimicrobial peptide), is also part of the cathelicidin family of host defense peptides. These include precursors of potent antimicrobial peptides that direct antimicrobial activity against various microbial pathogens and also activate mesenchymal cells during wound repair. CRAMP is expressed in testis, spleen, stomach and intestine.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CAMP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human