Anti-APOBR Rabbit Polyclonal Antibody (Cy7®)
Catalog # 10435-584
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein B Receptor
- Antigen Symbol:APOBR
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:55911
- Antigen Synonyms:APOB48R|APOB100R
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10435-584
- Supplier no.:BS-6336R-CY7
Specifications
About this item
APOB48R is a macrophage receptor that binds to the apolipoprotein B48 of dietary triglyceride (TG) rich lipoproteins. It may provide essential lipids, lipid soluble vitamins and other nutrients to reticuloendothelial cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APOBR
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat