Anti-Nucleoprotein TPR Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # 10422-796
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Nucleoprotein TPR
- Antigen Symbol:Nucleoprotein TPR
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:7175
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10422-796
- Supplier no.:BS-5755R-CY5.5
Specifications
About this item
Component of the cytoplasmic fibrils of the nuclear pore complex implicated in nuclear protein import. Its N-terminus is involved in activation of oncogenic kinases. Plays a role in the mitotic spindle checkpoint.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Nucleoprotein TPR
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat