Anti-OSGIN1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # 10422-052
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Oxidative Stress Induced Growth Inhibitor 1
- Antigen Symbol:OSGIN1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:29948
- Antigen Synonyms:huOKL38|Ovary|kidney and liver protein 38|OSGIN1|BDGI|Oxidative stress-induced growth inhibitor 1|OKL38|Pregnancy-induced growth inhibitor OKL38
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:380-430/560
- Purification:Purified by Protein A
- Cat. No.:10422-052
- Supplier no.:BS-5723R-A647
Specifications
About this item
Regulates the differentiation and proliferation of normal cells through the regulation of cell death.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: OSGIN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat