Anti-STAT5B Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10421-614
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Signal Transducer And Activator of Transcription 5B
- Antigen Symbol:STAT5B
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:6777
- Antigen Synonyms:STAT5|Signal transducer and activator of transcription 5B|Transcription factor STAT5B|Stat5b.|STA5B_HUMAN
- Modification:phospho-Ser731
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10421-614
- Supplier no.:BS-5703R-CY3
Specifications
About this item
Carries out a dual function: signal transduction and activation of transcription. Binds to the GAS element and activates PRL-induced transcription.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: STAT5B
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat