Order Entry
United States
ContactUsLinkComponent
 
Anti-EIF4B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  10413-788
 :  Bioss
Anti-EIF4B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  10413-788
 :  Bioss
 :  BS-5346R-FITC

 

  • Antibody Type:
    Primary
  • Antigen Name:
    eukaryotic translation initiation factor 4B
  • Antigen Symbol:
    EIF4B
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1975
  • Antigen Synonyms:
    EIF-4B|PRO1843
  • Modification:
    phospho-Ser504
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10413-788
  • Supplier no.:
    BS-5346R-FITC

 

 

eIF4B is required for the binding of mRNA to ribosomes. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F (referenced from swissprot).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EIF4B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human