Anti-EIF4B Rabbit Polyclonal Antibody (Cy7®)
Catalog # 10413-786
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:eukaryotic translation initiation factor 4B
- Antigen Symbol:EIF4B
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:1975
- Antigen Synonyms:EIF-4B|PRO1843
- Modification:phospho-Ser504
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10413-786
- Supplier no.:BS-5346R-CY7
Specifications
About this item
eIF4B is required for the binding of mRNA to ribosomes. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F (referenced from swissprot).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EIF4B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human