Anti-ERAS Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10405-150
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Embryonic stem cell-expressed Ras
- Antigen Symbol:ERAS
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:3266
- Antigen Synonyms:Embryonic stem cell-expressed Ras|ERAS|GTPase Eras|HRASP|E-Ras|HRAS2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10405-150
- Supplier no.:BS-4950R-CY5
Specifications
About this item
ERAS belongs to the small GTPase superfamily. Ras family. Ras proteins bind GDP/GTP and possess intrinsic GTPase activity. The ERas gene is expressed in embryonic stem (ES) cells and promotes their in vitro proliferation and tumorigenicity. ERas-null ES cells maintain pluripotency but show significantly reduced growth and tumorigenicity, which are rescued by expression of ERas complementary DNA or by activated phosphatidylinositol-3-OH kinase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ERAS
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat