Anti-AOC1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10405-696
: Bioss
- Antibody Type:Primary
- Antigen Name:Amine Oxidase, Copper Containing 1
- Antigen Symbol:AOC1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Plant
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:26
- Antigen Synonyms:ABP1|DAO|KAO|DAO1|ABP
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10405-696
- Supplier no.:BS-4731R-FITC
ABP1
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AOC1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant