Specifications
- Antibody Type:Primary
- Antigen Name:Defensin, beta 4A
- Antigen Symbol:DEFB4A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:1673
- Antigen Synonyms:DEFB2|BD-2|SAP1|DEFB4|DEFB-2|DEFB102|HBD-2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:34-64/64
- Purification:Purified by Protein A
- Cat. No.:10397-734
- Supplier no.:BS-4307R-A350
Specifications
About this item
Beta-defensins (also designated BD, and hBD in human) are small cationic peptides with broad-spectrum antimicrobial activity. Produced in mucosal epithelia and neutrophils of several species, Beta-defensins are developmentally regulated. Human b-defensin 2 is locally regulated by inflammation and is the first member of the b-defensin family that is locally inducible by inflammation. The murine homolog of human b-defensin 2, which is called b-defensin 3, is present in the respiratory system and in low levels in the epithelial cells of the intestine and lung. The unique murine b-defensin 2 (Defb2) is not expressed in airways of untreated mice, but is upregulated in the airways by lipopolysaccharide and may contribute to host defense at the mucosal surface of the airways
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DEFB4A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human