Order Entry
United States
ContactUsLinkComponent
Anti-PABP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-PABP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10388-532
Supplier:  Bioss
Anti-PABP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10388-532
Supplier:  Bioss
Supplier Number:  BS-3836R-FITC

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Poly(A)-binding protein
  • Antigen Symbol:
    PABP
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    26986
  • Antigen Synonyms:
    PABP|PABPC2|PABP1|PAB1|PABPL1
  • Modification:
    methyl-Arg455/460
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    440-480/636
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10388-532
  • Supplier no.:
    BS-3836R-FITC

Specifications

About this item

Binds the poly(A) tail of mRNA, including that of its own transcript. May be involved in cytoplasmic regulatory processes of mRNA metabolism such as pre-mRNA splicing. Its function in translational initiation regulation can either be enhanced by PAIP1 or repressed by PAIP2. Can probably bind to cytoplasmic RNA sequences other than poly(A) in vivo. Involved in translationally coupled mRNA turnover. Implicated with other RNA-binding proteins in the cytoplasmic deadenylation/translational and decay interplay of the FOS mRNA mediated by the major coding-region determinant of instability (mCRD) domain. Involved in regulation of nonsense-mediated decay (NMD) of mRNAs containing premature stop codons; for the recognition of premature termination codons (PTC) and initiation of NMD a competitive interaction between UPF1 and PABPC1 with the ribosome-bound release factors is proposed.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PABP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat