Anti-CRX Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # 10387-976
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Cone-rod Homeobox
- Antigen Symbol:CRX
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:1406
- Antigen Synonyms:LCA7|OTX3|CRD|CORD2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:60-110/299
- Purification:Purified by Protein A
- Cat. No.:10387-976
- Supplier no.:BS-3798R-CY5.5
Specifications
About this item
Transcription factor that binds and transactivates the sequence 5'-TAATC[CA]-3' which is found upstream of several photoreceptor-specific genes, including the opsin genes. Acts synergistically with other transcription factors, such as NRL, RORB and RAX, to regulate photoreceptor cell-specific gene transcription. Essential for the maintenance of mammalian photoreceptors.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CRX
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: