Order Entry
United States
ContactUsLinkComponent
Anti-SIAH1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-SIAH1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # 10371-166
Supplier:  Avantor
Anti-SIAH1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # 10371-166
Supplier:  Avantor

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    SIAH1/SIAH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    6478, 6477
  • Antigen Synonyms:
    Seven in Absentia Homolog 1/2
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    200-250/324
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10371-166
  • Supplier no.:
    BS-3634R-A488

Specifications

About this item

Siah2 is a E3 Ubiquitin ligase protein that mediates ubiquitination and subsequent proteasomal degradation of target proteins. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targets. Siah2 triggers the ubiquitin-mediated degradation of many proteins, including transcription regulators (e.g. POU2AF1, PML, NCOR1), cell surface receptors (e.g. DCC), antiapoptotic proteins (e.g. BAG1), and proteins involved in synaptic vesicle function in neurons (e.g. SYP). Siah2 has some overlapping function with SIAH1 but is able to trigger the ubiquitin-mediated degradation of TRAF2, whereas SIAH1 can not.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SIAH1/2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat