Anti-RPS6 Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10372-480
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Ribosomal Protein S6
- Antigen Symbol:RPS6
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:6194
- Antigen Synonyms:Phosphoprotein NP33|RPS6|40S ribosomal protein S6|S6|OK/SW-cl.2
- Modification:phospho-Ser240/Ser244
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:210-249/249
- Purification:Purified by Protein A
- Cat. No.:10372-480
- Supplier no.:BS-3389R-CY3
Specifications
About this item
May play an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RPS6
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat