Anti-RPS6 Rabbit Polyclonal Antibody (Cy7®)
10372-462
: Bioss
- Antibody Type:Primary
- Antigen Name:Ribosomal Protein S6
- Antigen Symbol:RPS6
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:6194
- Antigen Synonyms:Phosphoprotein NP33|RPS6|40S ribosomal protein S6|S6|OK/SW-cl.2
- Modification:phospho-Ser235/Ser236
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:200-245/249
- Purification:Purified by Protein A
- Cat. No.:10372-462
- Supplier no.:BS-3388R-CY7
May play an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RPS6
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat