Anti-IFNGR2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # 10346-186
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon gamma receptor 2
- Antigen Symbol:IFNGR2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:3460
- Antigen Synonyms:AF-1|IMD28|IFNGT1|IFGR2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:270-320/337
- Purification:Purified by Protein A
- Cat. No.:10346-186
- Supplier no.:BS-2710R-A488
Specifications
About this item
Part of the receptor for interferon gamma. Required for signal transduction. This accessory factor is an integral part of the IFN-gamma signal transduction pathway and is likely to interact with GAF, JAK1, and/or JAK2.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IFNGR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat