Order Entry
United States
ContactUsLinkComponent
Anti-LCK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-LCK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10343-632
Supplier:  Bioss
Anti-LCK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10343-632
Supplier:  Bioss
Supplier Number:  BS-2649R-FITC

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Lymphocyte-specific Protein tyrosine kinase
  • Antigen Symbol:
    LCK
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3932
  • Antigen Synonyms:
    p56lck|LSK|tyrosine-protein kinase Lck|YT16|leukocyte C-terminal Src kinase|T-lymphocyte specific protein tyrosine kinase p56lck|p56(LSTRA) protein-tyrosine kinase|RP4-675E8.4|t cell-specific protein-tyrosine kinase|lymphocyte cell-specific protein-tyrosine kinase|proto-oncogene tyrosine-protein kinase LCK|pp58lck
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10343-632
  • Supplier no.:
    BS-2649R-FITC

Specifications

About this item

Non-receptor tyrosine-protein kinase that plays an essential role in the selection and maturation of developing T-cells in the thymus and in the function of mature T-cells. Plays a key role in T-cell antigen receptor (TCR)-linked signal transduction pathways. Constitutively associated with the cytoplasmic portions of the CD4 and CD8 surface receptors. Association of the TCR with a peptide antigen-bound MHC complex facilitates the interaction of CD4 and CD8 with MHC class II and class I molecules, respectively, thereby recruiting the associated LCK protein to the vicinity of the TCR/CD3 complex. LCK then phosphorylates tyrosines residues within the immunoreceptor tyrosine-based activation motifs (ITAM) of the cytoplasmic tails of the TCR-gamma chains and CD3 subunits, initiating the TCR/CD3 signaling pathway. Once stimulated, the TCR recruits the tyrosine kinase ZAP7, that becomes phosphorylated and activated by LCK. Following this, a large number of signaling molecules are recruited, ultimately leading to lymphokine production. LCK also contributes to signaling by other receptor molecules. Associates directly with the cytoplasmic tail of CD2, which leads to hyperphosphorylation and activation of LCK. Also plays a role in the IL2 receptor-linked signaling pathway that controls the T-cell proliferative response. Binding of IL2 to its receptor results in increased activity of LCK. Is expressed at all stages of thymocyte development and is required for the regulation of maturation events that are governed by both pre-TCR and mature alpha beta TCR. Phosphorylates other substrates including RUNX3, PTK2B/PYK2, the microtubule-associated protein MAPT, RHOH or TYROBP.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LCK
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat