Anti-IL18RAP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10342-856
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Interleukin 18 Receptor Accessory Protein
- Antigen Symbol:IL18RAP
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:8807
- Antigen Synonyms:IL-18Rbeta|IL-1RAcPL|CDw218b|CD218b|IL18RB|IL-1R-7|IL-1R7|IL-18R-beta|ACPL|IL-18RAcP
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:20-50/599
- Purification:Purified by Protein A
- Cat. No.:10342-856
- Supplier no.:BS-2616R-FITC
Specifications
About this item
Required for the high affinity binding of interleukin 18 (IL-18) to its receptor complex (By similarity). Together with IL18R1 mediates IL-18-dependent activation of NF-kappa-B and JNK.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL18RAP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat