Anti-VCAN Rabbit Polyclonal Antibody
Catalog # 10341-302
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Versican
- Antigen Symbol:VCAN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:1462
- Antigen Synonyms:WGN1|PG-M|versican|GHAP|WGN|CSPG2|ERVR
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:3360-3395/3396
- Purification:Purified by Protein A
- Cat. No.:10341-302
- Supplier no.:BS-2533R
Specifications
About this item
May play a role in intercellular signaling and in connecting cells with the extracellular matrix. May take part in the regulation of cell motility, growth and differentiation. Binds hyaluronic acid.
Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: VCAN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat