Anti-EIF4E Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10337-514
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:eukaryotic translation initiation factor 4E
- Antigen Symbol:EIF4E
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:1977
- Antigen Synonyms:eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|eukaryotic translation initiation factor 4E-like 1|eIF-4E|CBP|eIF4E|EIF4F|mRNA cap-binding protein
- Modification:phospho-Ser209
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:200-217/217
- Purification:Purified by Protein A
- Cat. No.:10337-514
- Supplier no.:BS-2446R-CY5
Specifications
About this item
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures. Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit mediates the binding to the mRNA cap.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat