Anti-AMPH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10334-722
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Amphiphysin
- Antigen Symbol:AMPH
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:273
- Antigen Synonyms:HGNC:471|amphiphysin (Stiff-Man syndrome with breast cancer 128kDa autoantigen) |AMPH1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10334-722
- Supplier no.:BS-2409R-FITC
Specifications
About this item
Amphiphysin is associated with the cytoplasmic surface of synaptic vesicles. A subset of patients with stiff man syndrome who were also affected by breast cancer are positive for auto against this protein. Alternate splicing of this gene results in two transcript variants (Amphiphysin 1 and Amphiphysin 2) encoding different isoforms. Additional splice variants have been described, but their full length sequences have not been determined.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AMPH
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat