Anti-IL1RN Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # 10338-594
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Interleukin 1 Receptor Antagonist
- Antigen Symbol:IL1RN
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:16181
- Antigen Synonyms:IL1F3|IL-1ra|IL-1RN|DIRA|IRAP|IL-1ra3|ICIL-1RA|MVCD4|IL1RA
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:27-70/178
- Purification:Purified by Protein A
- Cat. No.:10338-594
- Supplier no.:BS-2216R-A647
Specifications
About this item
Inhibits the activity of interleukin-1 by binding to receptor IL1R1 and preventing its association with the coreceptor IL1RAP for signaling. Has no interleukin-1 like activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL1RN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat