Anti-ABCF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10333-916
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:ATP Binding Cassette, Subfamily F, Member 1
- Antigen Symbol:ABCF1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:23
- Antigen Synonyms:ATP-binding cassette sub-family F member 1|ATP-binding cassette 50|ABC50|TNF-alpha-stimulated ABC protein|ABCF1|ABC27
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:530-580/845
- Purification:Purified by Protein A
- Cat. No.:10333-916
- Supplier no.:BS-1960R-FITC
Specifications
About this item
Isoform 2 is required for efficient Cap- and IRES-mediated mRNA translation initiation. Isoform 2 is not involved in the ribosome biogenesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ABCF1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat