Specifications
- Antibody Type:Primary
- Antigen Name:Kh Domain Containing, Rna Binding, Signal Transduction Associated 1
- Antigen Symbol:KHDRBS1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10304-020
- Supplier no.:BS-13699R-FITC
Specifications
About this item
Sam 68 is a protein that is phosphorylated on tyrosine and functions as a substrate for Src family tyrosine kinases during mitosis. Sam 68 also associates with several SH2 and SH3 domain-containing signaling proteins, such as GRB2 and PLC ?. Originally cloned as Ras GAP-associated p62, further investigations have shown that Sam 68 and Ras GAP-associated p62 are not antigenically related, nor are they encoded by the same gene. Like Sam 68, the Sam 68-like mammalian proteins, SLM-1 and SLM-2, demonstrate RNA binding activity. Also like Sam 68, SLM-1 is tyrosine phosphorylated and functions as an adapter protein for signaling molecules, including GRB2, PLC ?, Fyn and Ras GAP. SLM-2 is not tyrosine phosphorylated, nor does it appear to associate with GRB2, PLC ?, Fyn or Ras GAP, indicating that SLM-2 may not be an adapter protein for these proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KHDRBS1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat