- Antibody Type:Primary
- Antigen Name:Deltex 2
- Antigen Symbol:DTX2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:113878
- Antigen Synonyms:Deltex homolog 2|RNF58
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10279-810
- Supplier no.:BS-12394R-HRP
DTX2 belongs to the Deltex family. It contains one RING-type zinc finger and two WWE domains. DTX2 is a regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. It probably acts both as a positive and negative regulator of Notch, depending on the developmental and cell context; mediates the antineural activity of Notch, possibly by inhibiting the transcriptional activation mediated by MATCH1. DTX2 also functions as an ubiquitin ligase protein in vitro, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity. The WWE domains are thought to mediate some protein-protein interaction, and are frequently found in ubiquitin ligases. There are two named isoforms.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: DTX2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat