Anti-SFRS7/9G8/SRp20 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10260-804
: Bioss
- Antibody Type:Primary
- Antigen Name:SFRS7/9G8/SRp20
- Antigen Symbol:SFRS7/9G8/SRp20
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:6432
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:1-50/238
- Purification:Purified by Protein A
- Cat. No.:10260-804
- Supplier no.:BS-11755R-FITC
Required for pre-mRNA splicing. Can also modulate alternative splicing in vitro. Represses the splicing of MAPT/Tau exon 10. May function as export adapter involved in mRNA nuclear export such as of histone H2A. Binds mRNA which is thought to be transferred to the NXF1-NXT1 heterodimer for export (TAP/NXF1 pathway); enhances NXF1-NXT1 RNA-binding activity. RNA-binding is semi-sequence specific.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SFRS7/9G8/SRp20
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat