Specifications
- Antibody Type:Primary
- Antigen Name:APH1B Gamma Secretase Subunit
- Antigen Symbol:APH1B
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:83464
- Antigen Synonyms:PSFL|PRO1328|APH-1B|TAAV688
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10263-732
- Supplier no.:BS-11640R-HRP
Specifications
About this item
Anterior pharynx defective 1 (Aph-1) is a polytopic, seven-pass membrane protein that functions as one of the four essential components in the presenilin-Gamma-secretase enzyme complex. This enzyme complex is necessary for the intra-membrane proteolysis of several different membrane proteins, including the beta-Amyloid precursor protein, and is involved in multiple neurodevelopmental signaling pathways. Aph-1b and Aph-1a are splice variants of Aph-1. Aph-1b specifically lacks exon 4, which encodes for the entire fourth transmembrane domain, causing the protein to be destabilized. Deficiency of Aph-1a causes a reduction in Gamma-secretase activity, however deficiency of Aph-1b does not; thus, Aph-1b may execute redundant functions in the cell. Aph-1b expression and Gamma-secretase activity may be implicated in neurodevelopmental disorders, such as schizophrenia.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: APH1B
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat