Specifications
- Antibody Type:Primary
- Antigen Name:N-Acetyltransferase 8B (Putative, Gene/Pseudogene)
- Antigen Symbol:NAT8B
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10265-496
- Supplier no.:BS-11592R-CY5
Specifications
About this item
Acetyltransferases and deacetylases are protein groups most often associated with oncogenesis and cell cycle regulation. NAT-8B (N-acetyltransferase 8B), also known as CML2 (camello-like protein 2), is a 227 amino acid single-pass membrane protein that is implicated in gastrulation regulation. A member of the camello family, NAT-8B contains one N-acetyltransferase domain and is encoded by a gene that maps to human chromosome 2p13.2. The NAT-8B gene is susceptible to a nonsense mutation at Serine 16, which leads to a stop codon and subsequently, a non-functional protein that is truncated in length. Similarly, a nonsense mutation at Glutamine 168 is thought to lead to a non-functional protein, as it causes the N-acetyltransferase to become disrupted. Human chromosome 2 consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstré°‰ syndrome.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NAT8B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat