To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cadherin Family Member 15
- Antigen Symbol:CDHF15
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Protocadherin Fat 3.|Cadherin family member 15|FAT3_HUMAN|FAT tumor suppressor homolog 3|Fat3|CDHR10|CDHF15|hFat3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10248-198
- Supplier no.:BS-11081R-A647
Specifications
About this item
The cadherins represent a family of Ca2+-dependent adhesion molecules that function to mediate cell to cell binding that is critical for the maintenance of structure and morphogenesis. Cadherins each contain a large extracellular domain at the N-terminus, which is characterized by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity. The relatively short C-terminal intracellular domain interacts with a variety of cytoplasmic proteins, including ∫-catenin, to regulate cadherin function. The cadherin superfamily includes cadherins, protocadherins, desmogleins and desmocollins. FAT3 (FAT tumor suppressor homolog 3, also known as CDHF15 or CDHR10, is a 4,589 amino acid single-pass type I membrane protein expressed in ES cells, primitive neuroectoderm, fetal brain, infant brain, adult neural tissues and prostate. Containing thirty-three cadherin domains, four EGF-like domains and one laminin G-like domain, FAT3 may participate in the interactions between neurites derived from specific subsets of neurons during development.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDHF15/FAT3
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: