Anti-Thyroid peroxidase Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10243-362
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Thyroid Peroxidase
- Antigen Symbol:TPO
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:7173
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:120-170/933
- Purification:Purified by Protein A
- Cat. No.:10243-362
- Supplier no.:BS-10406R-FITC
Specifications
About this item
Iodination and coupling of the hormonogenic tyrosines in thyroglobulin to yield the thyroid hormones T(3) and T(4).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Thyroid peroxidase
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat