Anti-HINT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10364-334
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Histidine triad nucleotide binding Protein 1
- Antigen Symbol:HINT1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:HINT1|PKCI1|Protein kinase C inhibitor 1|NMAN|Histidine triad nucleotide-binding protein 1|PRKCNH1|PKCI-1|HINT
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10364-334
- Supplier no.:BS-10228R-FITC
Specifications
About this item
The protein encoded by this gene can hydrolyze substrates such as AMP-morpholidate, AMP-N-alanine methyl ester, AMP-alpha-acetyl lysine methyl ester, and AMP-NH2. The encoded protein interacts with these substrates via a histidine triad motif, which is part of the loop that binds to the substrate. This gene has been found to be a tumor suppressing gene. Several transcript variants, but only one of them protein-coding, have been found for this gene. [provided by RefSeq, Dec 2012].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HINT1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat