Order Entry
United States
ContactUsLinkComponent
Anti-TSHR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-TSHR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10229-020
Supplier:  Avantor
Anti-TSHR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10229-020
Supplier:  Avantor

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Thyroid Stimulating Hormone Receptor
  • Antigen Symbol:
    TSHR
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    7253
  • Antigen Synonyms:
    hTSHR-I|CHNG1|LGR3
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    21-71/764
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10229-020
  • Supplier no.:
    BS-0003R-FITC

Specifications

About this item

Receptor for thyrothropin. Plays a central role in controlling thyroid cell metabolism. The activity of this receptor is mediated by G proteins which activate adenylate cyclase. Also acts as a receptor for thyrostimulin (GPA2+GPB5).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSHR
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat