ContactUsLinkComponent
Datalogging RH/Temperature Pen, Sper Scientific
Datalogging RH/Temperature Pen, Sper Scientific
Supplier:  Sper Scientific
Certificates
undefined
Datalogging RH/Temperature Pen, Sper Scientific
Supplier:  Sper Scientific
Specifications
  • Catalog No:
  • 10025-404
  • 10025-406
  • Interface:
  • USB
  • USB
  • Weight:
  • 34 g
  • 34 g
  • W×D×H:
  • 133×25 mm (5.25×1")
  • 133×25 mm (5.25×1")