QUANTISWAB® Irradiated Swabs for Media
95060-840
- Description:QUANTISWAB® Irradiated
- Size:200 swabs
- Cat. No.:95060-840
- Shelf life:18 months
- Supplier no.:43801
QUANTISWAB® is an irradiated, triple-wrapped environmental swab utilizing nylon-flocked technology. This patented technology offers the QUANTISWAB® a unique design allowing up to 60% recovery of micro-organisms from surfaces compared to a recovery rate of 20% for traditional swabs.
- Ideal for sampling of irregular surfaces
The QUANTISWAB® has also demonstrated a 90% release capacity of microorganisms captured by the swab.
Specifically designed for aseptic manufacturing facilities, QUANTISWAB® product is protected by triple sterile packaging, perfect for the biopharmaceutical and cosmetic industries.
: Made to order items have a lead time of 42 days.