General Purpose Viscosity Standards, Silicone Fluids, BrookField Engineering
Supplier: Avantor
Certificates
About this item
These viscosity standards is used to verify calibration of laboratory viscometers/rheometers.
- Accurate to ±1% of viscosity value
- Economical
- Excellent temperature stability
The selection of one or two fluids will normally provide sufficient measurement points to verify calibration of your instrument.
1 cP=1 mPa s
Certifications: NIST traceable certification
Frequently Bought Together